Learn More
Biomatik Corporation Mouse Interleukin 2,IL-2 ELISA kit
Supplier:  Biomatik Corporation EKC40078
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/ml - 2000 pg/ml. Sensitivity: 7.8 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04351. Target Synonyms: IL-2, TCGF, lymphokine, T cell growth factor|aldesleukin|interleukin-2|involved in regulation of T-cell clonal expansion. Restriction: For Research Use Only.
Catalog No. 50-228-7700