Learn More
Biomatik Corporation Mouse Neurotrophin 3, Nt-3 Elisa Kit
Supplier:  Biomatik Corporation EKC40106
Encompass_Preferred
Mouse Neurotrophin 3, Nt-3 Elisa Kit for detecting Neurotrophin 3, Nt-3 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20181; Ntf3. Target Synonyms: Ntf3; Ntf-3; Neurotrophin-3; NT-3; HDNF; Nerve growth factor 2; NGF-2; Neurotrophic factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7708