Learn More
Biomatik Corporation Human Vascular Endothelial Cell Growth Factor C, Vegf-C Elisa Kit
Supplier:  Biomatik Corporation EKC40129
Encompass_Preferred
Human Vascular Endothelial Cell Growth Factor C, Vegf-C Elisa Kit for detecting Vascular Endothelial Growth Factor C in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49767; VEGFC. Target Synonyms: Flt 4L; Flt4 ligand; FLT4 ligand DHM; Flt4-L; Flt4L; Vascular endothelial growth factor C; Vascular endothelial growth factor related protein; Vascular endothelial growth factor-related protein; VEGF C; VEGF-C; Vegfc; VEGFC_HUMAN; VRP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7710