Learn More
Biomatik Corporation Canine Testosterone (T) Elisa Kit
Supplier:  Biomatik Corporation EKC40243
Encompass_Preferred
Canine Testosterone (T) Elisa Kit for detecting Testosterone (T) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.1-20ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7716