Learn More
Biomatik Corporation Human Dihydrotestosterone, Dht Elisa Kit
Supplier:  Biomatik Corporation EKC40340
Encompass_Preferred
Human Dihydrotestosterone, Dht Elisa Kit for detecting Dihydrotestosterone, Dht in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 10-2000pg/ml. Sensitivity: 5pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7721