Learn More
Biomatik Corporation Mouse Tissue Inhibitors Of Metalloproteinase 1, Timp-1 Elisa Kit
Supplier:  Biomatik Corporation EKC40365
Encompass_Preferred
Mouse Tissue Inhibitors Of Metalloproteinase 1, Timp-1 Elisa Kit for detecting Timp Metallopeptidase Inhibitor 1 in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.110ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12032; Timp1. Target Synonyms: Timp1; Timp; Timp-1; Metalloproteinase inhibitor 1; Collagenase inhibitor 16C8 fibroblast; Erythroid-potentiating activity; EPA; TPA-S1; TPA-induced protein; Tissue inhibitor of metalloproteinases 1; TIMP-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7727