Learn More
Biomatik Corporation Mouse Angiotensinogen (Agt) Elisa Kit
Supplier:  Biomatik Corporation EKC40474
Encompass_Preferred
Mouse Angiotensinogen (Agt) Elisa Kit for detecting Angiotensinogen (Serpin Peptidase Inhibitor, Clade A, Member 8) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11859; Agt. Target Synonyms: Agt; Serpina8Angiotensinogen; Serpin A8) [Cleaved into: Angiotensin-1; Angiotensin 1-10; Angiotensin I; Ang I); Angiotensin-2; Angiotensin 1-8; Angiotensin II; Ang II); Angiotensin-3; Angiotensin 2-8; Angiotensin III; Ang III; Des-Asp[1]-angiotensin II); Angiotensin-4; Angiotensin 3-8; Angiotensin IV; Ang IV); Angiotensin 1-9; Angiotensin 1-7; Angiotensin 1-5; Angiotensin 1-4]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7739