Learn More
Biomatik Corporation Human Interferon Inducible T-Cell Chemoattractant, I-Tac Elisa Kit
Supplier:  Biomatik Corporation EKC40559
Encompass_Preferred
Human Interferon Inducible T-Cell Chemoattractant, I-Tac Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 11 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O14625; CXCL11. Target Synonyms: b R1; b-R1; Beta-R1; betaR1; C-X-C motif chemokine 11; Chemokine (C-X-C motif) ligand 11; Chemokine C-X-C motif ligand 11; CXC11; CXCL11; CXL11_HUMAN; H174; I TAC; I-TAC; Interferon gamma inducible protein 9; Interferon gamma-inducible protein 9; Interferon inducible T cell alpha chemoattractant; Interferon-inducible T-cell a chemoattractant I-TAC; Interferon-inducible T-cell alpha chemoattractant; IP 9; IP-9; IP9; ITAC; MGC102770; SCYB11; SCYB9B; Small inducible cytokine B11; Small inducible cytokine subfamily B (Cys-X-Cys) member 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7752