Learn More
Biomatik Corporation Human Galanin, Gal Elisa Kit
Supplier:  Biomatik Corporation EKC40561
Encompass_Preferred
Human Galanin, Gal Elisa Kit for detecting Galanin, Gal in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22466; GAL. Target Synonyms: GAL 1; GAL; GAL GMAP; GAL1; GALA_HUMAN; Galanin message associated peptide; Galanin message-associated peptide; Galanin precursor; Galanin prepropeptide; Galanin related peptide; Galanin/GMAP prepropeptide; GALN; GLNN; GMAP; MGC40167. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7754