Learn More
Biomatik Corporation Human Cytotoxic T Lymphocyte Associated Antigen 4, Ctla-4 Elisa Kit
Supplier:  Biomatik Corporation EKC40583
Encompass_Preferred
Human Cytotoxic T Lymphocyte Associated Antigen 4, Ctla-4 Elisa Kit for detecting Cytotoxic T-Lymphocyte-Associated Protein 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16410; CTLA4. Target Synonyms: CTLA4; CD152; Cytotoxic T-lymphocyte protein 4; Cytotoxic T-lymphocyte-associated antigen 4; CTLA-4; CD antigen CD152. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7758