Learn More
Biomatik Corporation Human Tnf Α Converting Enzyme, Tace Elisa Kit
Supplier:  Biomatik Corporation EKC40607
Encompass_Preferred
Human Tnf Α Converting Enzyme, Tace Elisa Kit for detecting Adam Metallopeptidase Domain 17 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P78536; ADAM17. Target Synonyms: ADAM17; CSVP; TACE; Disintegrin and metalloproteinase domain-containing protein 17; ADAM 17; Snake venom-like protease; TNF-alpha convertase; TNF-alpha-converting enzyme; CD antigen CD156b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7762