Learn More
Biomatik Corporation Pig Interferon Β, Ifn-Β/Ifnb Elisa Kit
Supplier:  Biomatik Corporation EKC40708
Encompass_Preferred
Pig Interferon Β, Ifn-Β/Ifnb Elisa Kit for detecting Interferon Β, Ifn-Β/Ifnb in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 50-2000pg/ml. Sensitivity: 25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q29412; IFNB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7771