Learn More
Biomatik Corporation Human Breast And Kidney Expressed Chemokine, Brak Elisa Kit
Supplier:  Biomatik Corporation EKC40727
Encompass_Preferred
Human Breast And Kidney Expressed Chemokine, Brak Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 14 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.39-25ng/ml. Sensitivity: 0.09ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95715; CXCL14. Target Synonyms: 1110031L23Rik; 1200006I23Rik; AI414372; BMAC; bolekine; BRAK; Breast and kidney; C-X-C motif chemokine 14; C-X-C motif chemokine ligand 14; Chaemokine; CXC motif; ligand 14; Chemokine (C-X-C motif) ligand 14; Chemokine BRAK; CXC chemokine in breast and kidney; CXCL14; CXL14_HUMAN; JSC; Kec; Kidney-expressed chemokine CXC; KS1; MGC10687; MGC124510; MGC90667; MIP 2 gamma; MIP-2G; MIP2G; MIP2gamma; NJAC; PRO273; PSEC0212; Scyb14; Small Inducible Cytokine B14; Small inducible cytokine subfamily B (Cys-X-Cys) member 14 (BRAK). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7775