Learn More
Biomatik Corporation Monkey Interleukin 6,IL-6 ELISA Kit
Supplier:  Biomatik Corporation EKC40735
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P51494. Target Synonyms: BSF2, HGF, HSF, IFNB2, IL-6, B cell stimulatory factor-2|B-cell differentiation factor|CTL differentiation factor|OTTHUMP00000158544|hybridoma growth factor|interleukin 6|interleukin BSF-2. Restriction: For Research Use Only.
Catalog No. 50-228-7777