Learn More
Biomatik Corporation Human Complement 1Q, C1Q Elisa Kit
Supplier:  Biomatik Corporation EKC40752
Encompass_Preferred
Human Complement 1Q, C1Q Elisa Kit for detecting Complement Component 1, Q Subcomponent, B Chain in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02746; C1QB. Target Synonyms: C1qb; C1QB_HUMAN; Complement C1q subcomponent subunit B; Complement component 1 q subcomponent B chain; Complement component 1 q subcomponent beta polypeptide; Complement component C1q B chain; Complement subcomponent C1q chain B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7779