Learn More
Biomatik Corporation Chicken Interferon Β, Ifn-Β/Ifnb Elisa Kit
Supplier:  Biomatik Corporation EKC40786
Encompass_Preferred
Chicken Interferon Β, Ifn-Β/Ifnb Elisa Kit for detecting Interferon Β, Ifn-Β/Ifnb in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 2.5-200pg/ml. Sensitivity: 3.125pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q90873; IFNB. Target Synonyms: IFNB; IFN2; Interferon type B; IFN-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7785