Learn More
Biomatik Corporation Human Urotensin Ⅱ ELISA kit
Supplier:  Biomatik Corporation EKC40843
Encompass_Preferred
Sample Type: serum, saliva, cerebrospinal fluid (CSF), tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O95399. Target Synonyms: PRO1068, U-II, UCN2, UII,Urotensin 2. Restriction: For Research Use Only.
Catalog No. 50-228-7790