Learn More
Biomatik Corporation Human Phospholipase A2 Receptor 1, 180Kda (Pla2R1) Elisa Kit
Supplier:  Biomatik Corporation EKC40948
Encompass_Preferred
Human Phospholipase A2 Receptor 1, 180Kda (Pla2R1) Elisa Kit for detecting Phospholipase A2 Receptor 1, 180Kda (Pla2R1) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.027-20ng/ml. Sensitivity: 0.027ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13018; PLA2R1. Target Synonyms: 180 kDa secretory phospholipase A2 receptor; C-type lectin domain family 13 member C; CLEC13C; M type receptor; M-type receptor; Phospholipase A2 receptor 1; PLA2 R; PLA2-R; PLA2G1R; PLA2IR; PLA2R; PLA2R_HUMAN; PLA2R1; Soluble PLA2-R; Soluble PLA2R; Soluble secretory phospholipase A2 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7802