Learn More
Biomatik Corporation Horse Progesterone(PROG) ELISA kit
Supplier:  Biomatik Corporation EKC40958
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Equus caballus (Horse). Detection Range: 0.25 ng/mL-100 ng/mL. Sensitivity: 0.25 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7805