Learn More
Biomatik Corporation Mouse Albumin (Alb)ELISA Kit
Supplier:  Biomatik Corporation EKC41072
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.781 μg/mL-200 μg/mL. Sensitivity: 0.110 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07724. Target Synonyms: DKFZp779N1935, PRO0883, PRO0903, PRO1341, cell growth inhibiting protein 42|growth-inhibiting protein 20|serum albumin. Restriction: For Research Use Only.
Catalog No. 50-228-7817