Learn More
Biomatik Corporation Rat Toll-Like Receptor 4 (Tlr4) Elisa Kit
Supplier:  Biomatik Corporation EKC41182
Encompass_Preferred
Rat Toll-Like Receptor 4 (Tlr4) Elisa Kit for detecting Toll-Like Receptor 4 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9QX05; Tlr4. Target Synonyms: Tlr4; Toll-like receptor 4; Toll4; CD antigen CD284. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7833