Learn More
Biomatik Corporation Human protein-arginine deiminase type-4(PADI4) ELISA kit
Supplier:  Biomatik Corporation EKC41208
Encompass_Preferred
Sample Type: serum, urine, cell culture supernates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9UM07. Target Synonyms: PAD, PAD4, PADI5, PDI4, PDI5, HL-60 PAD|PADI-H protein|peptidyl arginine deiminase, type V|protein-arginine deiminase. Restriction: For Research Use Only.
Catalog No. 50-228-7835