Learn More
Biomatik Corporation Human Alpha-enolase (ENO1/ENO1L1/MBPB1/MPB1) ELISA kit
Supplier:  Biomatik Corporation EKC41271
Encompass_Preferred
Sample Type: serum, tissue homogenates, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P06733. Target Synonyms: ENO1L1, MBP-1, MPB1, NNE, PPH, 2-phospho-D-glycerate hydro-lyase|MYC promoter-binding protein 1|alpha enolase like 1|enolase 1|non-neural enolase|phosphopyruvate hydratase|tau-crystallin. Restriction: For Research Use Only.
Catalog No. 50-228-7840