Learn More
Biomatik Corporation Mouse coagulation factor Ⅲ,FⅢ ELISA Kit
Supplier:  Biomatik Corporation EKC41314
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P20352. Target Synonyms: CD142, TF, TFA, coagulation factor III|tissue factor |coagulation factor 3,F3. Restriction: For Research Use Only.
Catalog No. 50-228-7843