Learn More
Biomatik Corporation Human Angiopoietin-Related Protein 4 (Angptl4) Elisa Kit
Supplier:  Biomatik Corporation EKC41357
Encompass_Preferred
Human Angiopoietin-Related Protein 4 (Angptl4) Elisa Kit for detecting Angiopoietin-Like 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9BY76; ANGPTL4. Target Synonyms: Angiopoietin like 4; Angiopoietin related protein 4; Angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGL4_HUMAN; ANGPT L2; ANGPT L4; ANGPTL2; Angptl4; ARP4; Fasting induced adipose factor; FIAF; HARP; Hepatic angiopoietin related protein; Hepatic fibrinogen/angiopoietin related protein; Hepatic fibrinogen/angiopoietin-related protein; HFARP; NL2; Peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin related protein; PGAR; pp1158; PPARG angiopoietin related protein; PSEC0166; TGQTL; UNQ171; Weakly similar to angiopoietin 1 [H.sapiens]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7847