Learn More
Biomatik Corporation Human lymphotactin,Lptn/LTN ELISA Kit
Supplier:  Biomatik Corporation EKC42270
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.39 ng/ml - 25 ng/ml. Sensitivity: 0.0975 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P47992. Target Synonyms: ATAC, LPTN, LTN, SCM-1, SCM-1a, SCM1, SCYC1, lymphotactin|small inducible cytokine subfamily C, member 1 (lymphotactin). Restriction: For Research Use Only.
Catalog No. 50-228-7857