Learn More
Biomatik Corporation Human Integrin-Linked Kinase 1, Ilk-1 Elisa Kit
Supplier:  Biomatik Corporation EKC42293
Encompass_Preferred
Human Integrin-Linked Kinase 1, Ilk-1 Elisa Kit for detecting Integrin-Linked Kinase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 39.06-2500pg/ml. Sensitivity: 9.77pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13418; ILK. Target Synonyms: 59 kDa serine/threonine protein kinase; 59 kDa serine/threonine-protein kinase; DKFZp686F1765; Epididymis secretory protein Li 28; HEL S 28; ILK 1; ILK 2; ILK; ILK-1; ILK-2; ILK_HUMAN; ILK1; ILK2; Integrin linked kinase 2; Integrin linked Kinase; Integrin linked protein kinase; Integrin-linked protein kinase; p59; p59ILK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7862