Learn More
Biomatik Corporation Mouse ribonuclease, RNase A family, 2 (liver, eosinophil-derived neurotoxin) (RNASE2) ELISA kit
Supplier:  Biomatik Corporation EKC42359
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O35291. Target Synonyms: EDN, RNS2. Restriction: For Research Use Only.
Catalog No. 50-228-7878