Learn More
Biomatik Corporation Mouse Cd81 Antigen (Cd81) Elisa Kit
Supplier:  Biomatik Corporation EKC42420
Encompass_Preferred
Mouse Cd81 Antigen (Cd81) Elisa Kit for detecting Cd81 Molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 47-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35762; Cd81. Target Synonyms: Cd81; Tapa1CD81 antigen; 26 kDa cell surface protein TAPA-1; Target of the antiproliferative 1; CD antigen CD81. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7883