Learn More
Biomatik Corporation Human ADM (Adrenomedullin) ELISA Kit
Supplier:  Biomatik Corporation EKF57231
Encompass_Preferred
Human ADM (Adrenomedullin) ELISA Kit for detecting ADM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 3.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35318. Target Synonyms: ADM; Adrenomedullin; AM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7894