Learn More
Biomatik Corporation Human CCK (Cholecystokinin) ELISA Kit
Supplier:  Biomatik Corporation EKF57284
Encompass_Preferred
Human CCK (Cholecystokinin) ELISA Kit for detecting CCK in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P06307. Target Synonyms: CCK; Cholecystokinin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7906