Learn More
Biomatik Corporation Human HSPA5 (78 kDa glucose-regulated protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57379
Encompass_Preferred
Human HSPA5 (78 kDa glucose-regulated protein) ELISA Kit for detecting HSPA5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P11021. Target Synonyms: GRP78; Glucose Regulated Protein 78; HSPA5; BIP; GRP-78; HSP70-5; MIF2; 78 kDa glucose-regulated protein; Endoplasmic reticulum lumenal Ca; 2+-binding protein grp78; Heat shock 70 kDa protein 5; heat shock 70kD protein 5; glucose-regulated protein; 78kD; Immunoglobulin heavy chain-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7931