Learn More
Biomatik Corporation Human NTN1 (Netrin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF57404
Encompass_Preferred
Human NTN1 (Netrin-1) ELISA Kit for detecting NTN1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O95631. Target Synonyms: Ntn1; Netrin 1; netrin-1; Lnetrin 1; Mouse; homolog of. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7936