Learn More
Biomatik Corporation Human ADAMTS5 (A disintegrin and metalloproteinase with thrombospondin motifs 5) ELISA Kit
Supplier:  Biomatik Corporation EKF57442
Encompass_Preferred
Human ADAMTS5 (A disintegrin and metalloproteinase with thrombospondin motifs 5) ELISA Kit for detecting ADAMTS5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UNA0. Target Synonyms: ADAMTS5; A disintegrin and metalloproteinase with thrombospondin motifs 5; ADAMTS11; ADMP-2; Aggrecanase 2; A disintegrin and metalloproteinase with thrombospondin motifs 11; a disintegrin-like and metalloprotease; reprolysin type with thrombospondintype 1 motif; 5; aggrecanase-2; ADAM metallopeptidase with thrombospondin type 1 motif; 5; ADAM-TS 11; ADAM-TS 5; ADAMTS-11; ADAM-TS5; ADAMTS-5; ADMP2; ADMP-2A disintegrin and metalloproteinase with thrombospondin motifs 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7942