Learn More
Biomatik Corporation Human TERT (Telomerase reverse transcriptase) ELISA Kit
Supplier:  Biomatik Corporation EKF57449
Encompass_Preferred
Human TERT (Telomerase reverse transcriptase) ELISA Kit for detecting TERT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O14746. Target Synonyms: TERT; Telomerase Reverse Transcriptase; CMM9; DKCA2; DKCB4; EST2; PFBMFT1; TCS1; TP2; TRT; Telomerase-associated protein 2; Telomerase catalytic subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7946