Learn More
Biomatik Corporation Human Aβ40 (Amyloid Beta 40) ELISA Kit
Supplier:  Biomatik Corporation EKF57545
Encompass_Preferred
Human Aβ40 (Amyloid Beta 40) ELISA Kit for detecting Aβ40 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05067. Target Synonyms: Aβ40; Amyloid Beta 40; Abeta 40; Beta-amyloid protein 40; Aβ; 1-40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7962