Learn More
Biomatik Corporation Bovine procalcitonin (PCT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31764
Encompass_Preferred
The Bovine procalcitonin (PCT) ELISA Kit allows for quantitative determination of procalcitonin (PCT) in serum, plasma, tissue Homogenates. Species Reactivity: Bovine. Detection Range: 39.06 pg/mL-2500 pg/mL. Sensitivity: 9.77 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-3hours. Restriction: For Research Use Only.
Catalog No. 50-149-7508