Learn More
Biomatik Corporation Human f-Hb (Free Haemoglobin) ELISA Kit
Supplier:  Biomatik Corporation EKF57609
Encompass_Preferred
Human f-Hb (Free Haemoglobin) ELISA Kit for detecting f-Hb in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P69905; P68871. Target Synonyms: f-Hb; Free Haemoglobin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7978