Learn More
Biomatik Corporation Human GABA (Gamma-aminobutyric acid) ELISA Kit
Supplier:  Biomatik Corporation EKF57616
Encompass_Preferred
Human GABA (Gamma-aminobutyric acid) ELISA Kit for detecting GABA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: GABA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7980