Learn More
Biomatik Corporation Human PDCD1LG1 (Programmed Cell Death Protein 1 Ligand 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57682
Encompass_Preferred
Human PDCD1LG1 (Programmed Cell Death Protein 1 Ligand 1) ELISA Kit for detecting PDCD1LG1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 3.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9NZQ7. Target Synonyms: PDCD1LG1; CD274; PDCD1L1; PDCD1LG1; PD-L1; B7-H; B7-H1; B7H1; CD274 antigen; CD274 molecule; B7 homolog 1; PDL1; PDCD1 ligand 1; programmed cell death 1 ligand 1; Programmed death ligand 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7985