Learn More
Biomatik Corporation Human DEFA1 (Defensin Alpha 1, Neutrophil) ELISA Kit
Supplier:  Biomatik Corporation EKF58001
Encompass_Preferred
Human DEFA1 (Defensin Alpha 1, Neutrophil) ELISA Kit for detecting DEFA1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P59665. Target Synonyms: DEFA1; DEF1; HNP-1; HP1; MRS; alpha 2; DEFA1B; DEFA2; defensin; alpha 1; myeloid-related sequence; defensin; alpha 1; HP-1; myeloid-related sequence; neutrophil defensin 1; alpha-Defensin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8003