Learn More
Biomatik Corporation Rat Copeptin ELISA Kit
Supplier:  Biomatik Corporation EKF58062
Encompass_Preferred
Rat Copeptin ELISA Kit for detecting CPP; Copeptin in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200pg/ml. Sensitivity: 1.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01186. Target Synonyms: CPP; Copeptin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8031