Learn More
Biomatik Corporation Human GAGs (Glycosaminoglycan) ELISA Kit
Supplier:  Biomatik Corporation EKF58110
Encompass_Preferred
Human GAGs (Glycosaminoglycan) ELISA Kit for detecting GAGs in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Mucopolysaccharide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8037