Learn More
Biomatik Corporation Human 3MH (3-methylhistidine) ELISA Kit
Supplier:  Biomatik Corporation EKF58146
Encompass_Preferred
Human 3MH (3-methylhistidine) ELISA Kit for detecting 3MH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400nmol/ml. Sensitivity: 3.75nmol/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: 3-methylhistidine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8046