Learn More
Biomatik Corporation Human BMP-2 (Bone Morphogenetic Protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58150
Encompass_Preferred
Human BMP-2 (Bone Morphogenetic Protein 2) ELISA Kit for detecting BMP-2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12643. Target Synonyms: BMP-2; Bone Morphogenetic Protein 2; BMP2A; BMP-2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8048