Learn More
Biomatik Corporation 5-HIAA (5-Hydroxyindoleacetic Acid) ELISA Kit
Supplier:  Biomatik Corporation EKF58183
Encompass_Preferred
5-HIAA (5-Hydroxyindoleacetic Acid) ELISA Kit for detecting 5-HIAA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: 5-HIAA; 5-Hydroxyindoleacetic Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8058