Learn More
Biomatik Corporation Mouse D2D (D-Dimer) ELISA Kit
Supplier:  Biomatik Corporation EKF58202
Encompass_Preferred
Mouse D2D (D-Dimer) ELISA Kit for detecting D2D in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500ng/ml. Sensitivity: 4.688ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: E9PV24; Q8K0E8; Q8VCM7. Target Synonyms: D-Dimer; D2D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8063