Learn More
Biomatik Corporation Human WNT7A (Protein Wnt-7a) ELISA Kit
Supplier:  Biomatik Corporation EKF58279
Encompass_Preferred
Human WNT7A (Protein Wnt-7a) ELISA Kit for detecting WNT7A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O00755. Target Synonyms: WNT7A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8084