Learn More
Biomatik Corporation MT (Melatonin) ELISA Kit
Supplier:  Biomatik Corporation EKF58332
Encompass_Preferred
MT (Melatonin) ELISA Kit for detecting MT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Melatonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8098