Learn More
Biomatik Corporation Human SAKL (Soluble alpha-Klotho) ELISA Kit
Supplier:  Biomatik Corporation EKF58413
Encompass_Preferred
Human SAKL (Soluble alpha-Klotho) ELISA Kit for detecting SAKL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UEF7. Target Synonyms: SAKL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8118